Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 12 | 2718 | ||
NM_001322855.1 | NP_001309784.1 | 13 | 2860 | |||
NM_001322856.1 | NP_001309785.1 | 13 | 2817 | |||
NM_001322857.1 | NP_001309786.1 | 12 | 2638 | |||
NM_001322858.1 | NP_001309787.1 | 13 | 2735 | |||
NM_001322859.1 | NP_001309788.1 | 10 | 2344 | |||
NM_001322860.1 | NP_001309789.1 | 9 | 2284 | |||
NM_001322861.1 | NP_001309790.1 | 9 | 1994 | |||
NM_001322862.1 | NP_001309791.1 | 13 | 2695 | |||
NM_001322863.1 | NP_001309792.1 | 12 | 2667 | |||
NM_001322864.1 | NP_001309793.1 | 11 | 2235 | |||
NM_001322866.1 | NP_001309795.1 | 12 | 2558 | |||
NM_001322868.1 | NP_001309797.1 | 10 | 2432 | |||
NM_020448.4 | NP_065181.1 | 12 | 2580 | |||
NR_136506.1 | 11 | 2488 | ||||
NR_136507.1 | 6 | 1995 | ||||
NR_136508.1 | 11 | 2449 | ||||
XM_011541808.2 | XP_011540110.1 | 11 | 2495 | |||
GenBank mRNA | BC063583.1 | - | 12 | 2510 | 4386115 | |
BX640883.1 | - | 13 | 2642 |