Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAMBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020799.3 | NP_065850.1 | 3 | 618 | ||
XM_006717928.2 | XP_006717991.1 | 4 | 719 | |||
XM_006717930.3 | XP_006717993.1 | 3 | 1206 | |||
XM_011539983.1 | XP_011538285.1 | 4 | 1126 | |||
XM_011539985.1 | XP_011538287.1 | 4 | 449 | |||
XM_017016453.1 | XP_016871942.1 | 4 | 1443 | |||
XM_017016454.1 | XP_016871943.1 | 5 | 927 | |||
XM_017016455.1 | XP_016871944.1 | 6 | 1382 | |||
XM_017016456.1 | XP_016871945.1 | 4 | 1039 | |||
XM_017016457.1 | XP_016871946.1 | 3 | 965 | |||
GenBank mRNA | AB010120.1 | - | 2 | 146 | ||
AB037794.1 | - | 3 | 939 | |||
AK056086.1 | - | 3 | 547 | |||
AK092946.1 | - | 2 | 747 | |||
BC010846.1 | - | 3 | 536 | 3898533 | ||
BX649096.1 | - | 3 | 8983 | |||
CD244526.1 | - | 3 | 643 |