Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267597.1 | NP_001254526.1 | 6 | 3369 | ||
NM_001267605.1 | NP_001254534.1 | 7 | 2029 | |||
NM_001267606.1 | NP_001254535.1 | 5 | 2778 | |||
NM_021045.2 | NP_066383.1 | 6 | 3741 | |||
XM_006717485.3 | XP_006717548.1 | 5 | 3698 | |||
XM_011519597.2 | XP_011517899.1 | 5 | 3290 | |||
XM_011519598.2 | XP_011517900.1 | 7 | 1679 | |||
XM_011519599.1 | XP_011517901.1 | 7 | 1632 | |||
XM_011519600.2 | XP_011517902.1 | 6 | 1520 | |||
XM_017016444.1 | XP_016871933.1 | 4 | 3347 | |||
XM_017016445.1 | XP_016871934.1 | 5 | 3838 | |||
XM_017016446.1 | XP_016871935.1 | 6 | 3389 | |||
XM_017016447.1 | XP_016871936.1 | 5 | 1635 | |||
XM_017016450.1 | XP_016871939.1 | 6 | 1971 | |||
GenBank mRNA | AJ491695.1 | - | 6 | 3663 | ||
AL049341.1 | - | 1 | 24 | |||
BC110869.1 | - | 5 | 2568 | |||
BC132915.1 | - | 6 | 3399 | 40146937 | ||
BC132917.1 | - | 6 | 3399 | 40146939 |