Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267597.1 | NP_001254526.1 | 6 | 2927 | ||
NM_001267605.1 | NP_001254534.1 | 7 | 1587 | |||
NM_001267606.1 | NP_001254535.1 | 5 | 2336 | |||
NM_021045.2 | NP_066383.1 | 6 | 3299 | |||
XM_006717485.3 | XP_006717548.1 | 5 | 3256 | |||
XM_011519597.2 | XP_011517899.1 | 5 | 2848 | |||
XM_011519598.2 | XP_011517900.1 | 7 | 1237 | |||
XM_011519599.1 | XP_011517901.1 | 7 | 1190 | |||
XM_011519600.2 | XP_011517902.1 | 6 | 1078 | |||
XM_017016444.1 | XP_016871933.1 | 4 | 2905 | |||
XM_017016445.1 | XP_016871934.1 | 5 | 3396 | |||
XM_017016446.1 | XP_016871935.1 | 6 | 2947 | |||
XM_017016447.1 | XP_016871936.1 | 5 | 1193 | |||
XM_017016450.1 | XP_016871939.1 | 6 | 1529 | |||
GenBank mRNA | AJ491695.1 | - | 6 | 3221 | ||
BC110869.1 | - | 5 | 2126 | |||
BC132915.1 | - | 6 | 2957 | 40146937 | ||
BC132917.1 | - | 6 | 2957 | 40146939 |