Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002809.3 | NP_002800.2 | 10 | 1594 | ||
GenBank mRNA | AF091075.1 | - | 3 | 303 | ||
AK022896.1 | - | 10 | 1505 | |||
AK054611.1 | - | 10 | 1538 | |||
AK055799.1 | - | 11 | 1475 | |||
AK094206.1 | - | 11 | 1516 | |||
AK293539.1 | - | 11 | 1480 | |||
AK298411.1 | - | 10 | 1433 | |||
AK300081.1 | - | 10 | 1261 | |||
AK300973.1 | - | 11 | 1510 | |||
BC000074.2 | - | 10 | 1494 | 3505074 | ||
BC004859.2 | - | 10 | 1456 | 3832823 | ||
BC012302.1 | - | 10 | 1542 | 3868681 | ||
BC020518.1 | - | 10 | 1492 | 3901555 | ||
BC025686.1 | - | 10 | 1492 | 5205360 | ||
BT007217.1 | - | 10 | 1391 | |||
CR456772.1 | - | 10 | 1391 | |||
D67025.1 | - | 10 | 1548 |