Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001156474.1 | NP_001149946.1 | 13 | 2005 | ||
NM_021827.4 | NP_068599.3 | 12 | 1735 | |||
XM_005274161.1 | XP_005274218.1 | 12 | 1861 | |||
XM_005274162.3 | XP_005274219.1 | 8 | 906 | |||
XM_017018105.1 | XP_016873594.1 | 11 | 1359 | |||
GenBank mRNA | AK027167.1 | - | 8 | 1033 | ||
AK131331.1 | - | 12 | 1738 | |||
AK223168.1 | - | 8 | 1039 | |||
AK298875.1 | - | 9 | 1054 | |||
BC126412.1 | - | 13 | 1849 | 8992128 |