Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM111A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142519.2 | NP_001135991.1 | 5 | 649 | ||
NM_001142520.2 | NP_001135992.1 | 5 | 672 | |||
NM_001142521.2 | NP_001135993.1 | 5 | 485 | |||
NM_001312909.1 | NP_001299838.1 | 6 | 887 | |||
NM_001312910.1 | NP_001299839.1 | 5 | 711 | |||
NM_001312911.1 | NP_001299840.1 | 4 | 476 | |||
NM_022074.3 | NP_071357.2 | 4 | 383 | |||
NM_198847.2 | NP_942144.1 | 4 | 344 | |||
GenBank mRNA | AB067482.1 | - | 2 | 787 | ||
AK092061.1 | - | 4 | 327 | |||
AK092237.1 | - | 5 | 2977 | |||
AK291453.1 | - | 4 | 349 | |||
BC071759.1 | - | 4 | 343 | 6451741 | ||
BQ420366.1 | - | 4 | 476 | |||
BQ948815.1 | - | 5 | 572 | |||
CR749358.1 | - | 3 | 990 | |||
DA502464.1 | - | 4 | 383 |