Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002812.4 | NP_002803.2 | 7 | 1138 | ||
GenBank mRNA | AK301771.1 | - | 7 | 1149 | ||
BC001164.1 | - | 7 | 889 | 3528096 | ||
BC065006.1 | - | 7 | 877 | 6055235 | ||
D38047.1 | - | 7 | 877 | |||
EU668354.1 | - | 7 | 1138 |