Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002814.3 | NP_002805.1 | 5 | 715 | ||
NM_170750.2 | NP_736606.1 | 5 | 636 | |||
GenBank mRNA | AB009619.1 | - | 5 | 613 | ||
AK058108.1 | - | 5 | 689 | |||
AK295996.1 | - | 5 | 530 | |||
AY057056.1 | - | 5 | 534 | |||
BC011960.1 | - | 5 | 624 | 3881328 | ||
BQ217943.1 | - | 5 | 631 | |||
D83197.1 | - | 5 | 689 |