Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022786.1 | NP_073623.1 | 4, 5 | 700 | ||
XM_017002106.1 | XP_016857595.1 | 5, 6 | 794 | |||
GenBank mRNA | AF271780.1 | - | 4, 5 | 700 | ||
AF290878.1 | - | 4, 5 | 700 | |||
AF321442.1 | - | 4, 5 | 734 | |||
AK293049.1 | - | 4, 5 | 706 | |||
AL122047.1 | - | 1, 2 | 99 | |||
BC016309.1 | - | 4, 5 | 695 | 4047121 |