Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324475.1 | NP_001311404.1 | 2 | 354 | ||
NM_023015.4 | NP_075391.3 | 1 | 695 | |||
XM_005245459.2 | XP_005245516.1 | 1 | 712 | |||
XM_006711490.3 | XP_006711553.1 | 1 | 55 | |||
GenBank mRNA | AK290025.1 | - | 2 | 348 | ||
AL832133.1 | - | 1 | 79 | |||
BC105092.1 | - | 1 | 158 | 8144095 | ||
BC105094.1 | - | 1 | 158 | 8144097 | ||
BC116458.1 | - | 1 | 690 | 5752962 | ||
BX640786.1 | - | 1 | 681 | |||
BX640950.1 | - | 1 | 696 | |||
CR627233.1 | - | 1 | 695 | |||
CR749212.1 | - | 1 | 689 | |||
CR749376.1 | - | 1 | 696 |