Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHACTR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048183.1 | NP_001041648.1 | 5 | 581 | ||
NM_023923.3 | NP_076412.3 | 4 | 564 | |||
XM_005245970.2 | XP_005246027.1 | 5 | 588 | |||
XM_017002157.1 | XP_016857646.1 | 5 | 588 | |||
XM_017002158.1 | XP_016857647.1 | 6 | 498 | |||
XM_017002159.1 | XP_016857648.1 | 6 | 747 | |||
XM_017002160.1 | XP_016857649.1 | 4 | 564 | |||
XM_017002161.1 | XP_016857650.1 | 6 | 578 | |||
XM_017002162.1 | XP_016857651.1 | 7 | 640 | |||
XM_017002163.1 | XP_016857652.1 | 5 | 1193 | |||
XM_017002164.1 | XP_016857653.1 | 5 | 578 | |||
XM_017002165.1 | XP_016857654.1 | 6 | 551 | |||
XM_017002166.1 | XP_016857655.1 | 6 | 578 | |||
XM_017002167.1 | XP_016857656.1 | 7 | 640 | |||
XM_017002168.1 | XP_016857657.1 | 6 | 551 | |||
XM_017002169.1 | XP_016857658.1 | 6 | 481 | |||
XM_017002170.1 | XP_016857659.1 | 4 | 622 | |||
XM_017002171.1 | XP_016857660.1 | 7 | 715 | |||
XM_017002172.1 | XP_016857661.1 | 4 | 399 | |||
XM_017002173.1 | XP_016857662.1 | 6 | 481 | |||
GenBank mRNA | AK025436.1 | - | 6 | 568 | ||
BC021247.2 | - | 4 | 543 | 4577873 | ||
BC029266.1 | - | 5 | 440 | 4422413 | ||
BC068508.1 | - | 6 | 449 | 4825777 | ||
CR749449.1 | - | 5 | 560 |