Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142674.1 | NP_001136146.1 | 14 | 2762 | ||
NM_001142675.1 | NP_001136147.1 | 13 | 2872 | |||
NM_001142676.1 | NP_001136148.1 | 14 | 2733 | |||
NM_001142677.1 | NP_001136149.1 | 12 | 2565 | |||
NM_023947.3 | NP_076436.3 | 13 | 2658 | |||
GenBank mRNA | AK027711.1 | - | 13 | 2607 | ||
AK124697.1 | - | 13 | 3234 | |||
BC095409.1 | - | 13 | 2594 | 30333427 |