Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099285.1 | NP_001092755.1 | 5 | 1074 | ||
NM_002823.4 | NP_002814.3 | 5 | 1071 | |||
XM_011511518.1 | XP_011509820.1 | 5 | 1804 | |||
XM_011511519.1 | XP_011509821.1 | 5 | 1801 | |||
XM_011511520.1 | XP_011509822.1 | 5 | 1703 | |||
XM_011511521.1 | XP_011509823.1 | 5 | 1700 | |||
XM_011511522.2 | XP_011509824.1 | 4 | 2564 | |||
GenBank mRNA | AK223605.1 | - | 5 | 1066 | ||
BC003510.1 | - | 5 | 1054 | 3610808 | ||
BC013859.1 | - | 1 | 366 | |||
BC022433.1 | - | 5 | 1070 | 4277490 | ||
BC034921.1 | - | 5 | 1062 | 4335961 | ||
BC066905.1 | - | 5 | 992 | 4830132 | ||
BC071647.1 | - | 5 | 1049 | 5112610 | ||
BC071879.1 | - | 5 | 1051 | 6425103 | ||
BC098420.1 | - | 5 | 1056 | 5017164 | ||
BX647606.1 | - | 1 | 2485 | |||
M14630.1 | - | 5 | 1071 | |||
M26708.1 | - | 5 | 1043 |