Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024522.2 | NP_078798.2 | 7 | 1817 | ||
XM_017002320.1 | XP_016857809.1 | 6 | 1411 | |||
GenBank mRNA | AK022712.1 | - | 6 | 1389 | ||
AK298874.1 | - | 5 | 1309 | |||
AL832547.1 | - | 4 | 1082 |