Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301039.1 | NP_001287968.1 | 5 | 351 | ||
NM_001301040.1 | NP_001287969.1 | 5 | 351 | |||
NM_001301041.1 | NP_001287970.1 | 3 | 137 | |||
NM_001301042.1 | NP_001287971.1 | 4 | 221 | |||
NM_024579.3 | NP_078855.2 | 4 | 240 | |||
XM_011509984.2 | XP_011508286.1 | 4 | 267 | |||
GenBank mRNA | AA095731.1 | - | 4 | 208 | ||
AK026874.1 | - | 4 | 225 | |||
AX770997.1 | - | 4 | 238 | |||
AX770998.1 | - | 5 | 265 | |||
AX770999.1 | - | 4 | 240 | |||
BC017761.1 | - | 4 | 225 | 4617936 | ||
DA358536.1 | - | 3 | 148 |