Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSANTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308027.1 | NP_001294956.1 | 4 | 1010 | ||
NM_001312919.1 | NP_001299848.1 | 3 | 754 | |||
NM_001312920.1 | NP_001299849.1 | 3 | 1744 | |||
NM_001312921.1 | NP_001299850.1 | 4 | 867 | |||
NM_024631.3 | NP_078907.2 | 4 | 854 | |||
XM_006718905.1 | XP_006718968.1 | 4 | 2379 | |||
XM_006718906.3 | XP_006718969.1 | 4 | 2215 | |||
XM_011542981.2 | XP_011541283.2 | 5 | 1568 | |||
XM_017018295.1 | XP_016873784.1 | 4 | 17625 | |||
XM_017018296.1 | XP_016873785.1 | 4 | 2535 | |||
XM_017018297.1 | XP_016873786.1 | 5 | 836 | |||
XM_017018298.1 | XP_016873787.1 | 5 | 13596 | |||
XM_017018299.1 | XP_016873788.1 | 5 | 637 | |||
GenBank mRNA | AK026995.1 | - | 4 | 853 | ||
AK092413.1 | - | 3 | 1418 | |||
AK303902.1 | - | 3 | 733 | |||
AL512691.1 | - | 3 | 1744 | |||
BC064919.1 | - | 4 | 995 | 6093009 |