Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024654.4 | NP_078930.3 | 8 | 1415 | ||
XM_005263493.3 | XP_005263550.1 | 6 | 747 | |||
XM_011542147.2 | XP_011540449.1 | 3 | 341 | |||
XM_017002338.1 | XP_016857827.1 | 6 | 925 | |||
XM_017002339.1 | XP_016857828.1 | 5 | 641 | |||
GenBank mRNA | AK026976.1 | - | 4 | 425 | ||
AK091284.1 | - | 8 | 1352 | |||
AK289653.1 | - | 8 | 1347 | |||
BC009257.2 | - | 5 | 505 | |||
BC063639.1 | - | 5 | 455 | |||
BC073860.1 | - | 4 | 418 | |||
BC094836.1 | - | 7 | 790 | |||
BC105095.1 | - | 8 | 1415 | 8144098 | ||
BC110849.1 | - | 8 | 1324 | 6167575 | ||
BC112278.1 | - | 8 | 1415 | 8327746 | ||
BC143646.1 | - | 8 | 1415 |