Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144030.1 | NP_001137502.1 | 3 | 419 | ||
NM_024662.2 | NP_078938.2 | 5 | 634 | |||
GenBank mRNA | AB051496.1 | - | 5 | 555 | ||
AK001636.1 | - | 5 | 489 | |||
AK022241.1 | - | 5 | 596 | |||
AK074430.1 | - | 1212 | ||||
AK225341.1 | - | 5 | 596 | |||
AK294044.1 | - | 3 | 318 | |||
AL136882.1 | - | 5 | 491 | |||
BC020205.1 | - | 5 | 456 | |||
BC035558.1 | - | 5 | 568 | 5166101 | ||
CN336530.1 | - | 5 | 636 |