Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144030.1 | NP_001137502.1 | 12, 13 | 1547 | ||
NM_024662.2 | NP_078938.2 | 14, 15 | 1762 | |||
GenBank mRNA | AB051496.1 | - | 14, 15 | 1683 | ||
AF489535.1 | - | 10, 11 | 1059 | |||
AK001636.1 | - | 14, 15 | 1617 | |||
AK022241.1 | - | 14, 15 | 1724 | |||
AK225341.1 | - | 14, 15 | 1724 | |||
AK294044.1 | - | 12, 13 | 1446 | |||
AL136882.1 | - | 14, 15 | 1619 | |||
BC020205.1 | - | 14, 15 | 1584 | |||
BC035558.1 | - | 14, 15 | 1696 | 5166101 |