Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136474.2 | NP_001129946.1 | 9 | 1552 | ||
NM_001136475.2 | NP_001129947.1 | 7 | 1268 | |||
NM_001301056.1 | NP_001287985.1 | 8 | 1748 | |||
NM_024749.4 | NP_079025.2 | 6 | 1616 | |||
XM_011509987.2 | XP_011508289.1 | 11 | 2032 | |||
XM_011509988.2 | XP_011508290.1 | 12 | 2106 | |||
XM_011509991.2 | XP_011508293.2 | 7 | 1445 | |||
XM_011509994.1 | XP_011508296.1 | 5 | 1318 | |||
XM_011509995.1 | XP_011508297.1 | 6 | 1148 | |||
XM_017002351.1 | XP_016857840.1 | 9 | 1866 | |||
XM_017002352.1 | XP_016857841.1 | 6 | 1299 | |||
GenBank mRNA | AK022567.1 | - | 9 | 1478 | ||
AK096605.1 | - | 9 | 1416 | |||
AK302675.1 | - | 7 | 1121 | |||
BC051856.1 | - | 6 | 1501 | 6067012 |