Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184917.2 | NP_001171846.1 | 9, 10 | 943 | ||
NM_001256433.2 | NP_001243362.1 | 7, 8 | 727 | |||
NM_001256434.2 | NP_001243363.1 | 9, 10 | 941 | |||
NM_001256435.2 | NP_001243364.1 | 8, 9 | 1173 | |||
NM_001282203.1 | NP_001269132.1 | 8, 9 | 1301 | |||
NM_001282204.1 | NP_001269133.1 | 8, 9 | 1017 | |||
NM_002861.4 | NP_002852.1 | 8, 9 | 889 | |||
XM_005256386.3 | XP_005256443.1 | 9, 10 | 1093 | |||
XM_005256387.3 | XP_005256444.1 | 9, 10 | 1013 | |||
XM_006722287.3 | XP_006722350.1 | 9, 10 | 868 | |||
XM_017024910.1 | XP_016880399.1 | 8, 9 | 814 | |||
XM_017024911.1 | XP_016880400.1 | 8, 9 | 959 | |||
XM_017024912.1 | XP_016880401.1 | 9, 10 | 1065 | |||
GenBank mRNA | AK093328.1 | - | 8, 9 | 1237 | ||
AK297996.1 | - | 8, 9 | 950 | |||
AK301723.1 | - | 8, 9 | 1051 | |||
AK315777.1 | - | 9, 10 | 865 | |||
AK316385.1 | - | 8, 9 | 710 | |||
AY251539.1 | - | 9, 10 | 834 | |||
BC000351.2 | - | 8, 9 | 797 | 2961418 | ||
BC010075.2 | - | 8, 9 | 840 | 4303545 | ||
BX419403.2 | - | 7, 8 | 667 | |||
CR456779.1 | - | 8, 9 | 767 | |||
D84307.1 | - | 8, 9 | 833 |