Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024763.4 | NP_079039.4 | 4 | 671 | ||
NM_207014.2 | NP_996897.2 | 4 | 671 | |||
XM_005271212.3 | XP_005271269.1 | 4 | 624 | |||
XM_011542161.2 | XP_011540463.1 | 4 | 624 | |||
XM_011542162.2 | XP_011540464.1 | 4 | 624 | |||
XM_017002353.1 | XP_016857842.1 | 4 | 624 | |||
XM_017002354.1 | XP_016857843.1 | 4 | 624 | |||
XM_017002356.1 | XP_016857845.1 | 3 | 445 | |||
XM_017002357.1 | XP_016857846.1 | 4 | 624 | |||
GenBank mRNA | AK127011.1 | - | 3 | 373 | ||
AK292830.1 | - | 5 | 624 | |||
AL516734.3 | - | 4 | 618 | |||
BC032406.1 | - | 4 | 570 | |||
BC126379.1 | - | 4 | 646 | 8992095 | ||
BC126381.1 | - | 4 | 646 | 8992097 | ||
BX648840.1 | - | 4 | 672 |