Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136040.2 | NP_001129512.1 | 4 | 433 | ||
NM_001142565.1 | NP_001136037.1 | 4 | 433 | |||
NM_024811.3 | NP_079087.3 | 4 | 509 | |||
XM_005274298.4 | XP_005274355.1 | 4 | 519 | |||
XM_005274299.4 | XP_005274356.1 | 4 | 436 | |||
XM_005274303.4 | XP_005274360.1 | 5 | 464 | |||
XM_011545257.2 | XP_011543559.1 | 4 | 519 | |||
XM_011545258.2 | XP_011543560.1 | 4 | 390 | |||
XM_011545259.2 | XP_011543561.1 | 4 | 390 | |||
XM_011545260.2 | XP_011543562.1 | 5 | 510 | |||
XM_011545261.2 | XP_011543563.1 | 5 | 511 | |||
XM_011545262.2 | XP_011543564.1 | 5 | 457 | |||
XM_011545263.2 | XP_011543565.1 | 5 | 456 | |||
XM_017018345.1 | XP_016873834.1 | 4 | 519 | |||
GenBank mRNA | AJ275970.1 | - | 3 | 299 | ||
AK092037.1 | - | 4 | 440 | |||
AK096343.1 | - | 4 | 389 | |||
AK294578.1 | - | 4 | 389 | |||
AL832546.1 | - | 2 | 104 | |||
AL832667.1 | - | 4 | 424 | |||
AL833332.1 | - | 4 | 389 | |||
BC018135.1 | - | 4 | 399 | 3913173 | ||
BX537888.1 | - | 4 | 414 | |||
DA194038.1 | - | 4 | 509 |