Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM110D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024869.2 | NP_079145.2 | 2 | 1336 | ||
GenBank mRNA | AK024112.1 | - | 2 | 1325 | ||
BC025658.1 | - | 2 | 1322 | 5198480 |