Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271833.1 | NP_001258762.1 | 2 | 400 | ||
NM_024901.4 | NP_079177.2 | 2 | 468 | |||
XM_006710921.2 | XP_006710984.2 | 3 | 422 | |||
XM_006710922.1 | XP_006710985.1 | 2 | 412 | |||
XM_006710923.1 | XP_006710986.1 | 2 | 400 | |||
XM_006710924.1 | XP_006710987.1 | 2 | 177 | |||
XM_011542187.1 | XP_011540489.1 | 3 | 433 | |||
XM_011542189.1 | XP_011540491.1 | 2 | 427 | |||
XM_011542190.2 | XP_011540492.1 | 3 | 301 | |||
XM_017002386.1 | XP_016857875.1 | 2 | 177 | |||
XM_017002387.1 | XP_016857876.1 | 2 | 193 | |||
XM_017002388.1 | XP_016857877.1 | 2 | 427 | |||
XM_017002389.1 | XP_016857878.1 | 3 | 422 | |||
GenBank mRNA | AK026110.1 | - | 2 | 271 | ||
AL713773.1 | - | 2 | 311 | |||
BC004557.2 | - | 2 | 424 | 3957641 | ||
BP270686.1 | - | 2 | 400 |