Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271833.1 | NP_001258762.1 | 4 | 579 | ||
NM_024901.4 | NP_079177.2 | 4 | 647 | |||
XM_006710921.2 | XP_006710984.2 | 5 | 601 | |||
XM_006710922.1 | XP_006710985.1 | 4 | 591 | |||
XM_006710923.1 | XP_006710986.1 | 4 | 579 | |||
XM_006710924.1 | XP_006710987.1 | 4 | 356 | |||
XM_011542187.1 | XP_011540489.1 | 5 | 612 | |||
XM_011542189.1 | XP_011540491.1 | 4 | 606 | |||
XM_011542190.2 | XP_011540492.1 | 5 | 480 | |||
XM_017002386.1 | XP_016857875.1 | 4 | 356 | |||
XM_017002387.1 | XP_016857876.1 | 4 | 372 | |||
XM_017002388.1 | XP_016857877.1 | 4 | 606 | |||
XM_017002389.1 | XP_016857878.1 | 5 | 601 | |||
GenBank mRNA | AK026110.1 | - | 4 | 450 | ||
AK301583.1 | - | 3 | 443 | |||
AL713773.1 | - | 4 | 490 | |||
BC004557.2 | - | 4 | 603 | 3957641 |