Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002292.3 | NP_001002292.3 | 11 | 1729 | ||
NM_001193334.1 | NP_001180263.1 | 10 | 1462 | |||
NM_024911.6 | NP_079187.3 | 11 | 1790 | |||
XM_011542191.2 | XP_011540493.1 | 11 | 1713 | |||
XM_011542192.2 | XP_011540494.1 | 11 | 1723 | |||
XM_017002390.1 | XP_016857879.1 | 11 | 1557 | |||
GenBank mRNA | AK026744.1 | - | 8 | 871 | ||
AK074984.1 | - | 11 | 1712 | |||
AK301613.1 | - | 10 | 1322 | |||
AK309779.1 | - | 12 | 1669 | |||
AY359035.1 | - | 11 | 1772 | |||
BC007211.1 | - | 10 | 1328 | |||
BC110826.1 | - | 11 | 1691 | 6183768 | ||
BC137109.1 | - | 11 | 1587 | 9021106 | ||
BC137113.1 | - | 11 | 1587 | 9021110 | ||
BX538320.1 | - | 11 | 1729 | |||
BX648748.1 | - | 10 | 1460 | |||
DQ323735.1 | - | 11 | 1759 |