Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024625.1 | - | 3, 4 | 329 | ||
AK302075.1 | - | 1, 2 | 96 | |||
AL832217.1 | - | 6, 7 | 927 | |||
AY639646.1 | - | 2, 3 | 410 | |||
DC397297.1 | - | 1, 2 | 96 |