Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243718.1 | NP_001230647.1 | 5 | 1444 | ||
NM_001243719.1 | NP_001230648.1 | 8 | 1211 | |||
NM_002869.4 | NP_002860.2 | 8 | 1756 | |||
NM_198896.1 | NP_942599.1 | 8 | 1756 | |||
GenBank mRNA | AF052130.1 | - | 1 | 420 | ||
AK057157.1 | - | 8 | 1721 | |||
AK290132.1 | - | 8 | 1405 | |||
AK299639.1 | - | 6 | 1081 | |||
BC003617.2 | - | 8 | 1652 | 3506585 | ||
BC068486.1 | - | 9 | 1697 | 30342562 | ||
BC096818.1 | - | 8 | 1690 | 5295149 |