Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323550.1 | NP_001310479.1 | 4 | 884 | ||
NM_001323551.1 | NP_001310480.1 | 4 | 887 | |||
NM_025079.2 | NP_079355.2 | 4 | 887 | |||
XM_011542198.2 | XP_011540500.1 | 4 | 887 | |||
GenBank mRNA | AK026884.1 | - | 4 | 853 | ||
AK093343.1 | - | 4 | 679 | |||
AY920403.1 | - | 3 | 737 | |||
BC005001.1 | - | 4 | 811 | 2958199 |