Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320143.1 | NP_001307072.1 | 5, 6 | 1067 | ||
NM_001320144.1 | NP_001307073.1 | 5, 6 | 928 | |||
NM_025228.3 | NP_079504.2 | 5, 6 | 988 | |||
NR_109871.1 | 5, 6 | 988 | ||||
XM_011510018.2 | XP_011508320.1 | 5, 6 | 972 | |||
XM_011510019.1 | XP_011508321.1 | 5, 6 | 972 | |||
XM_017002399.1 | XP_016857888.1 | 5, 6 | 972 | |||
XM_017002400.1 | XP_016857889.1 | 5, 6 | 972 | |||
GenBank mRNA | AK122792.1 | - | 5, 6 | 971 | ||
AK125601.1 | - | 5, 6 | 1067 | |||
AK225762.1 | - | 5, 6 | 883 | |||
AK300555.1 | - | 6, 7 | 833 | |||
AK312755.1 | - | 5, 6 | 882 | |||
AL049667.1 | - | 5, 6 | 781 | |||
BC110302.1 | - | 5, 6 | 954 | 5219542 | ||
BC130417.1 | - | 4, 5 | 668 | 40146448 | ||
BC144139.1 | - | 4, 5 | 668 | 9052659 | ||
BX648450.1 | - | 5, 6 | 967 |