Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322462.1 | NP_001309391.1 | 9 | 3192 | ||
NM_001322463.1 | NP_001309392.1 | 7 | 2567 | |||
NM_001322464.1 | NP_001309393.1 | 7 | 2934 | |||
NM_030645.1 | NP_085148.1 | 7 | 2521 | |||
XM_017002404.1 | XP_016857893.1 | 6 | 2724 | |||
XM_017002405.1 | XP_016857894.1 | 6 | 2503 | |||
XM_017002406.1 | XP_016857895.1 | 8 | 2477 | |||
XM_017002407.1 | XP_016857896.1 | 6 | 2409 | |||
GenBank mRNA | AB051507.1 | - | 6 | 2228 | ||
AK056345.1 | - | 4 | 1706 | |||
AK056382.1 | - | 7 | 2521 | |||
AK091164.1 | - | 2 | 2443 | |||
AK125138.1 | - | 7 | 3198 | |||
AK125837.1 | - | 9 | 3461 | |||
AL136569.1 | - | 6 | 2108 | |||
AY358453.1 | - | 7 | 2443 | |||
BC010871.1 | - | 4 | 1730 | |||
BC011962.1 | - | 1 | 1160 | |||
BC017254.2 | - | 6 | 2010 | 3051344 |