Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258392.2 | NP_001245321.1 | 7 | 1151 | ||
NM_001258393.2 | NP_001245322.1 | 6 | 1064 | |||
NM_001258394.2 | NP_001245323.1 | 9 | 1348 | |||
NM_030813.5 | NP_110440.1 | 8 | 1241 | |||
XM_005274320.1 | XP_005274377.1 | 7 | 1027 | |||
XM_011545289.2 | XP_011543591.1 | 8 | 1114 | |||
GenBank mRNA | AK023214.1 | - | 7 | 1001 | ||
AK302006.1 | - | 6 | 907 | |||
AK302069.1 | - | 9 | 1046 | |||
AK302158.1 | - | 6 | 982 | |||
AL136909.1 | - | 8 | 1140 | |||
BC006404.2 | - | 8 | 1081 | 4304523 |