Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258392.2 | NP_001245321.1 | 12 | 1569 | ||
NM_001258393.2 | NP_001245322.1 | 11 | 1482 | |||
NM_001258394.2 | NP_001245323.1 | 14 | 1766 | |||
NM_030813.5 | NP_110440.1 | 13 | 1659 | |||
XM_005274320.1 | XP_005274377.1 | 12 | 1445 | |||
GenBank mRNA | AK023214.1 | - | 12 | 1419 | ||
AK302006.1 | - | 11 | 1325 | |||
AK302069.1 | - | 14 | 1464 | |||
AK302158.1 | - | 11 | 1400 | |||
AL136909.1 | - | 13 | 1558 | |||
AL834484.1 | - | 4 | 294 | |||
BC006257.2 | - | 5 | 376 | |||
BC006404.2 | - | 13 | 1499 | 4304523 |