Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030816.4 | NP_110443.3 | 4 | 968 | ||
XM_005271234.3 | XP_005271291.1 | 4 | 1211 | |||
XM_005271235.3 | XP_005271292.1 | 4 | 958 | |||
XM_006710929.3 | XP_006710992.1 | 4 | 981 | |||
XM_017002413.1 | XP_016857902.1 | 2 | 191 | |||
XM_017002414.1 | XP_016857903.1 | 4 | 319 | |||
XM_017002415.1 | XP_016857904.1 | 4 | 319 | |||
GenBank mRNA | AL136717.1 | - | 3 | 864 | ||
BC028840.1 | - | 4 | 957 | 4558748 |