Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030971.3 | NP_112233.2 | 12 | 1595 | ||
GenBank mRNA | AK074707.1 | - | 11 | 1208 | ||
AK093011.1 | - | 3 | 1047 | |||
AK299405.1 | - | 11 | 1214 | |||
BC000124.1 | - | 11 | 1201 | 3352015 |