Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030971.3 | NP_112233.2 | 12 | 3064 | ||
GenBank mRNA | AK074707.1 | - | 11 | 2674 | ||
AK093011.1 | - | 3 | 2515 | |||
BC000124.1 | - | 11 | 2670 | 3352015 |