Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010984.3 | NP_001010984.1 | 2, 3 | 765 | ||
NM_032018.6 | NP_114407.3 | 2, 3 | 765 | |||
GenBank mRNA | AK027613.1 | - | 2, 3 | 445 | ||
AL512744.1 | - | 2, 3 | 379 | |||
AY358611.1 | - | 2, 3 | 1598 | |||
BC015740.1 | - | 2, 3 | 653 | |||
BC068478.1 | - | 2, 3 | 716 | 5298048 |