Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010984.3 | NP_001010984.1 | 4 | 2546 | ||
NM_001261462.2 | NP_001248391.1 | 3 | 2417 | |||
NM_032018.6 | NP_114407.3 | 5 | 1508 | |||
XM_006711818.2 | XP_006711881.1 | 4 | 1351 | |||
XM_011544289.2 | XP_011542591.1 | 3 | 834 | |||
GenBank mRNA | AK027613.1 | - | 5 | 1188 | ||
AL512744.1 | - | 5 | 1122 | |||
AY358611.1 | - | 4 | 3379 | |||
BC068478.1 | - | 5 | 1459 | 5298048 |