Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170631.1 | NP_001164102.1 | 6 | 1461 | ||
XM_011510051.2 | XP_011508353.1 | 7 | 1410 | |||
XM_011510052.2 | XP_011508354.1 | 6 | 1171 | |||
XM_017002442.1 | XP_016857931.1 | 7 | 1406 | |||
GenBank mRNA | AF354295.1 | - | 4 | 910 | ||
AK298394.1 | - | 6 | 1437 | |||
AY063125.1 | - | 4 | 844 |