Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032125.2 | NP_115501.2 | 6 | 1113 | ||
NR_037576.1 | 7 | 1266 | ||||
NR_037577.1 | 7 | 1201 | ||||
NR_037580.1 | 8 | 1354 | ||||
GenBank mRNA | AK095889.1 | - | 2 | 2342 | ||
AK123218.1 | - | 3 | 2361 | |||
AK222642.1 | - | 6 | 992 | |||
AK303821.1 | - | 6 | 1067 | |||
AL136683.1 | - | 6 | 1027 | |||
BC011579.1 | - | 6 | 1010 | |||
BC090039.1 | - | 7 | 1158 | 6181849 |