Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098794.1 | NP_001092264.1 | 11 | 2837 | ||
NM_032127.3 | NP_115503.2 | 11 | 2879 | |||
XM_006718343.2 | XP_006718406.1 | 11 | 2801 | |||
XM_011520397.2 | XP_011518699.1 | 11 | 2815 | |||
XM_011520398.2 | XP_011518700.1 | 11 | 2860 | |||
XM_011520399.2 | XP_011518701.1 | 11 | 2883 | |||
GenBank mRNA | AB051546.1 | - | 11 | 2742 | ||
AL136723.1 | - | 11 | 2813 | |||
BC030825.1 | - | 11 | 2750 | 4811942 |