Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199829.1 | NP_001186758.1 | 7 | 470 | ||
NM_032132.4 | NP_115508.2 | 8 | 491 | |||
XM_006711570.2 | XP_006711633.1 | 7 | 385 | |||
XM_006711571.2 | XP_006711634.1 | 4 | 247 | |||
XM_011510053.2 | XP_011508355.1 | 8 | 559 | |||
XM_011510054.2 | XP_011508356.1 | 8 | 513 | |||
XM_011510055.2 | XP_011508357.1 | 7 | 309 | |||
XM_011510056.2 | XP_011508358.1 | 3 | 91 | |||
GenBank mRNA | AK097390.1 | - | 8 | 479 | ||
AK098633.1 | - | 8 | 449 | |||
AL136755.2 | - | 7 | 467 | |||
AY626344.1 | - | 7 | 467 | |||
BC033014.1 | - | 6 | 392 | 4827705 | ||
BC047406.1 | - | 8 | 479 | 5298160 | ||
CR533505.1 | - | 6 | 352 |