Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199829.1 | NP_001186758.1 | 14 | 1394 | ||
NM_032132.4 | NP_115508.2 | 15 | 1415 | |||
XM_006711570.2 | XP_006711633.1 | 14 | 1309 | |||
XM_006711571.2 | XP_006711634.1 | 11 | 1171 | |||
XM_011510053.2 | XP_011508355.1 | 15 | 1483 | |||
XM_011510055.2 | XP_011508357.1 | 14 | 1233 | |||
XM_011510056.2 | XP_011508358.1 | 10 | 1015 | |||
GenBank mRNA | AK097390.1 | - | 15 | 1403 | ||
AK098633.1 | - | 15 | 1373 | |||
AL136755.2 | - | 14 | 1391 | |||
AY626344.1 | - | 14 | 1391 | |||
BC033014.1 | - | 13 | 1316 | 4827705 | ||
BC047406.1 | - | 15 | 1403 | 5298160 |