Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC88B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032251.5 | NP_115627.6 | 17 | 2981 | ||
XM_006718519.3 | XP_006718582.1 | 17 | 2993 | |||
XM_011544957.2 | XP_011543259.1 | 17 | 2994 | |||
XM_011544959.2 | XP_011543261.1 | 17 | 2994 | |||
XM_011544962.2 | XP_011543264.1 | 4 | 616 | |||
GenBank mRNA | AK074126.1 | - | 11 | 2415 | ||
AK090436.1 | - | 17 | 2976 | |||
AY491401.1 | - | 1 | 56 | |||
BC040232.1 | - | 4 | 538 | |||
BC140822.1 | - | 17 | 2970 | 9021690 | ||
BC140824.1 | - | 17 | 2970 | 9021692 | ||
BC141866.1 | - | 17 | 2960 |