Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032270.4 | NP_115646.2 | 3 | 958 | ||
XM_006710960.3 | XP_006711023.1 | 3 | 2600 | |||
XM_011542282.2 | XP_011540584.1 | 3 | 945 | |||
XM_011542283.2 | XP_011540585.1 | 4 | 2512 | |||
XM_017002503.1 | XP_016857992.1 | 3 | 805 | |||
XM_017002504.1 | XP_016857993.1 | 4 | 714 | |||
GenBank mRNA | AB081509.1 | - | 3 | 839 | ||
AK127881.1 | - | 3 | 895 | |||
AL136919.1 | - | 1 | 458 | |||
BC113973.1 | - | 3 | 765 | 40007964 |