Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297741.1 | NP_001284670.1 | 2 | 327 | ||
NM_001297742.1 | NP_001284671.1 | 2 | 517 | |||
NM_032320.6 | NP_115696.2 | 3 | 485 | |||
XM_017018405.1 | XP_016873894.1 | 2 | 1496 | |||
XM_017018406.1 | XP_016873895.1 | 3 | 475 | |||
XM_017018407.1 | XP_016873896.1 | 3 | 455 | |||
XM_017018408.1 | XP_016873897.1 | 3 | 455 | |||
XM_017018409.1 | XP_016873898.1 | 2 | 322 | |||
XM_017018410.1 | XP_016873899.1 | 2 | 314 | |||
XM_017018411.1 | XP_016873900.1 | 2 | 314 | |||
GenBank mRNA | AF329102.2 | - | 3 | 485 | ||
AK095946.1 | - | 3 | 414 | |||
AK292392.1 | - | 3 | 410 | |||
AK294259.1 | - | 2 | 306 | |||
AK294768.1 | - | 2 | 232 | |||
AK298226.1 | - | 2 | 202 | |||
AK310935.1 | - | 2 | 210 | |||
AK316163.1 | - | 2 | 517 | |||
AL832981.1 | - | 3 | 466 | |||
AY221959.2 | - | 3 | 528 | |||
BC005071.1 | - | 3 | 447 | 3531347 |