Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032323.2 | NP_115699.1 | 4 | 2048 | ||
NR_026678.1 | 4 | 2073 | ||||
GenBank mRNA | AK056816.1 | - | 3 | 2289 | ||
AK122634.1 | - | 5 | 1794 | |||
BC005094.1 | - | 4 | 2038 | 3955668 |