Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143943.1 | NP_001137415.1 | 6 | 446 | ||
NM_001290327.1 | NP_001277256.1 | 7 | 558 | |||
NM_032328.3 | NP_115704.1 | 7 | 621 | |||
NR_026587.1 | 5 | 361 | ||||
XM_011544300.2 | XP_011542602.1 | 7 | 580 | |||
XM_011544301.1 | XP_011542603.1 | 8 | 863 | |||
XM_011544302.2 | XP_011542604.1 | 6 | 442 | |||
XM_017002538.1 | XP_016858027.1 | 6 | 443 | |||
XM_017002539.1 | XP_016858028.1 | 6 | 443 | |||
XM_017002540.1 | XP_016858029.1 | 6 | 443 | |||
XM_017002541.1 | XP_016858030.1 | 6 | 443 | |||
XM_017002542.1 | XP_016858031.1 | 6 | 443 | |||
XM_017002543.1 | XP_016858032.1 | 6 | 443 | |||
XM_017002544.1 | XP_016858033.1 | 6 | 429 | |||
GenBank mRNA | AB209286.1 | - | 6 | 454 | ||
AK001393.1 | - | 5 | 300 | |||
AK090927.1 | - | 7 | 905 | |||
AK302883.1 | - | 6 | 389 | |||
BC005357.1 | - | 6 | 385 | 3511019 | ||
BG546306.1 | - | 7 | 492 | |||
BG701985.1 | - | 7 | 625 | |||
BI838837.1 | - | 7 | 509 | |||
BQ438351.1 | - | 6 | 388 | |||
DB286332.1 | - | 6 | 448 |